Skip to Content
Merck
o-Dianisidine dihydrochloride tablet, 10 mg substrate per tablet

o-Dianisidine dihydrochloride

o-Dianisidine dihydrochloride tablet, 10 mg substrate per tablet
Synonym(s):
3,3′-Dimethoxybenzidine dihydrochloride, Fast Blue B
Empirical Formula (Hill Notation):
C14H16N2O2 · 2HCl
CAS Number:
Molecular Weight:
317.21
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-3 of 3