Norfloxacin impurity A
Synonym(s):
7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Empirical Formula (Hill Notation):
C12H9ClFNO3
CAS Number:
Molecular Weight:
269.66
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing