Nω-Nitro-L-arginine methyl ester hydrochloride
Synonym(s):
L-NAME hydrochloride
Empirical Formula (Hill Notation):
C7H15N5O4 · HCl
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing