![N4-Acetylcytosine 99%](/deepweb/assets/sigmaaldrich/product/structures/612/809/497acf05-d40e-475a-94b6-7e66b25d5124/640/497acf05-d40e-475a-94b6-7e66b25d5124.png)
N4-Acetylcytosine
![N4-Acetylcytosine 99%](/deepweb/assets/sigmaaldrich/product/structures/612/809/497acf05-d40e-475a-94b6-7e66b25d5124/640/497acf05-d40e-475a-94b6-7e66b25d5124.png)
Empirical Formula (Hill Notation):
C6H7N3O2
CAS Number:
Molecular Weight:
153.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing