![MRQ30 Clear CD Vial™, Blue cap, Pre-slit TEF/Silicone septa volume 1.2 mL, clear glass vial, thread for 9 mm, O.D. × H × I.D. 12 mm × 32 mm × 6 mm, pkg of 100 ea](/deepweb/assets/sigmaaldrich/product/images/284/923/028fae46-92f0-4364-a7e7-57347ca28b0d/640/028fae46-92f0-4364-a7e7-57347ca28b0d.jpg)
MRQ30 Clear CD Vial™, Blue cap, Pre-slit TEF/Silicone septa
![MRQ30 Clear CD Vial™, Blue cap, Pre-slit TEF/Silicone septa volume 1.2 mL, clear glass vial, thread for 9 mm, O.D. × H × I.D. 12 mm × 32 mm × 6 mm, pkg of 100 ea](/deepweb/assets/sigmaaldrich/product/images/284/923/028fae46-92f0-4364-a7e7-57347ca28b0d/640/028fae46-92f0-4364-a7e7-57347ca28b0d.jpg)
Synonym(s):
Certified CD™ Vial (Center Draining) Kit, 9 mm thread with cap/septa, unassembled
Products
Product Category
Brand
ID (mm)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Showing 1-1 of 1