![Morantel Tartrate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/361/229/050a0ab5-6c57-4d05-88b6-a58585efdc9b/640/050a0ab5-6c57-4d05-88b6-a58585efdc9b.png)
Morantel tartrate
![Morantel Tartrate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/361/229/050a0ab5-6c57-4d05-88b6-a58585efdc9b/640/050a0ab5-6c57-4d05-88b6-a58585efdc9b.png)
Synonym(s):
Morantel (+)-tartrate salt, 1,4,5,6-Tetrahydro-1-methyl-2-(2-[3-methyl-2-thienyl]ethenyl)pyrimidine
Empirical Formula (Hill Notation):
C12H16N2S · C4H6O6
CAS Number:
Molecular Weight:
370.42
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2