Skip to Content
Merck
Mitotane United States Pharmacopeia (USP) Reference Standard

Mitotane

Mitotane United States Pharmacopeia (USP) Reference Standard
Synonym(s):
1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane, (2,4′-Dichlorodiphenyl)dichloroethane, o,p′-DDD, Mitotane, 1-Chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethyl]-benzene, 2,4′-DDD, o,p′-DDD
Linear Formula:
(ClC6H4)2CHCHCl2
CAS Number:
Molecular Weight:
320.04

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade