Skip to Content
Merck
Minoxidil Related Compound E pharmaceutical secondary standard, certified reference material

Minoxidil Related Compound E

Minoxidil Related Compound E pharmaceutical secondary standard, certified reference material
Synonym(s):
Mitoxantrone dihydrochloride, 1,4-Dihydroxy-5,8-bis[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione dihydrochloride
Empirical Formula (Hill Notation):
C22H28N4O6 · 2HCl
CAS Number:
Molecular Weight:
517.40

Products

Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1