![Methysticin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/565/065/f75b7932-61e1-492f-9264-2a2be0f2b28a/640/f75b7932-61e1-492f-9264-2a2be0f2b28a.png)
Methysticin
![Methysticin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/565/065/f75b7932-61e1-492f-9264-2a2be0f2b28a/640/f75b7932-61e1-492f-9264-2a2be0f2b28a.png)
Synonym(s):
R-(+)-Methysticin, (R)-6-[(E)-2-(1,3-Benzodioxol-5-yl)ethenyl]-5,6-dihydro-4-methoxy-2H-pyran-2-one, d-Methysticin, Kavahin, Kavatin
Empirical Formula (Hill Notation):
C15H14O5
CAS Number:
Molecular Weight:
274.27
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing