Methyl decanoate
Synonym(s):
Capric acid methyl ester, Decanoic acid methyl ester, Methyl caprate, Methyl decanoate
Linear Formula:
CH3(CH2)8COOCH3
CAS Number:
Molecular Weight:
186.29
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4