Hydroxysafflor yellow A
Synonym(s):
2,4-Di-β-D-glucopyranosyl-3,4,5-trihydroxy-6-[(2E)-3-(4-hydroxyphenyl)-1-oxo-2-propen-1-yl]-2,5-cyclohexadien-1-one, HSYA, Safflomin A
Empirical Formula (Hill Notation):
C27H32O16
CAS Number:
Molecular Weight:
612.53
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing