![Glyphosate-2-13C,15N PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/112/221/9da46033-8fde-4a16-a1cb-5afd476333d3/640/9da46033-8fde-4a16-a1cb-5afd476333d3.png)
Glyphosate-2-13C,15N
![Glyphosate-2-13C,15N PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/112/221/9da46033-8fde-4a16-a1cb-5afd476333d3/640/9da46033-8fde-4a16-a1cb-5afd476333d3.png)
Linear Formula:
HO2C13CH215NHCH2PO3H2
CAS Number:
Molecular Weight:
171.06
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing