
Glycidol

Synonym(s):
(±)-Glycidol, (±)-Oxirane-2-methanol, 2,3-Epoxy-1-propanol, Glycerolglycide
Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1