![Glutardialdehyde (50% solution in water) for synthesis](/deepweb/assets/sigmaaldrich/product/structures/194/500/db30dba8-9c07-4485-a2b8-113363fde2e9/640/db30dba8-9c07-4485-a2b8-113363fde2e9.png)
Glutardialdehyde
![Glutardialdehyde (50% solution in water) for synthesis](/deepweb/assets/sigmaaldrich/product/structures/194/500/db30dba8-9c07-4485-a2b8-113363fde2e9/640/db30dba8-9c07-4485-a2b8-113363fde2e9.png)
Synonym(s):
Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial
Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Products
Brand
Formula Weight
Physical Form
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing