![γ-Linolenic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/112/719/26f01dfc-cc65-47e2-a6e7-6e7745b554f7/640/26f01dfc-cc65-47e2-a6e7-6e7745b554f7.png)
γ-Linolenic acid
![γ-Linolenic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/112/719/26f01dfc-cc65-47e2-a6e7-6e7745b554f7/640/26f01dfc-cc65-47e2-a6e7-6e7745b554f7.png)
Synonym(s):
cis,cis,cis-6,9,12-Octadecatrienoic acid
Empirical Formula (Hill Notation):
C18H30O2
CAS Number:
Molecular Weight:
278.43
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Format
Tissue
Quality Segment
Functional Group
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing