Skip to Content
Merck
Ginsenoside Rc phyproof® Reference Substance

Ginsenoside Rc

Ginsenoside Rc phyproof® Reference Substance
Synonym(s):
(3β,12β)-20-[(6-O-α-L-Arabinofuranosyl-β-D-glucopyranosyl)oxy]-12-hydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, NSC 310104, Panaxoside Rc
Empirical Formula (Hill Notation):
C53H90O22
CAS Number:
Molecular Weight:
1079.27

Products

Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing