Skip to Content
Merck
Fluticasone propionate for system suitability CRS, European Pharmacopoeia (EP) Reference Standard

Fluticasone propionate for system suitability

Fluticasone propionate for system suitability CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Fluticasone propionate, (6α,11β,16α,17α)-6,9-Difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)androsta-1,4-diene-17-carbothioic acid S-(fluoromethyl) ester
Empirical Formula (Hill Notation):
C25H31F3O5S
CAS Number:
Molecular Weight:
500.57

Products

Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1