Skip to Content
Merck
Exo-Phenyl Kwon [2.2.1] Bicyclic Phosphine 95% (HPLC)

Exo-Phenyl Kwon [2.2.1] Bicyclic Phosphine

Exo-Phenyl Kwon [2.2.1] Bicyclic Phosphine 95% (HPLC)
Synonym(s):
(1S,4S,5R)-5-(phenyl)-2-tosyl-2-aza-5-phosphabicyclo[2.2.1]heptane
Empirical Formula (Hill Notation):
C18H20NO2PS
Molecular Weight:
345.40

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing