Ethyl 4-hydroxybenzoate sodium salt
Synonym(s):
Ethyl 4-hydroxybenzoate sodium salt
Empirical Formula (Hill Notation):
C9H9NaO3
CAS Number:
Molecular Weight:
188.16
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing