Skip to Content
Merck
Ethyl 2-chloro-2-(hydroxyimino)acetate 97%

Ethyl 2-chloro-2-(hydroxyimino)acetate

Ethyl 2-chloro-2-(hydroxyimino)acetate 97%
Synonym(s):
1-Ethyl oxalyl chloride 2-oxime
Linear Formula:
HON=C(Cl)CO2C2H5
CAS Number:
Molecular Weight:
151.55

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing