![Drometrizole United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/101/221/5e3ec369-8045-42d8-ba91-724c9bd7fe47/640/5e3ec369-8045-42d8-ba91-724c9bd7fe47.png)
Drometrizole
![Drometrizole United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/101/221/5e3ec369-8045-42d8-ba91-724c9bd7fe47/640/5e3ec369-8045-42d8-ba91-724c9bd7fe47.png)
Synonym(s):
2-(2-Hydroxy-5-methylphenyl)benzotriazole, 2-(2H-Benzotriazol-2-yl)-p-cresol
Empirical Formula (Hill Notation):
C13H11N3O
CAS Number:
Molecular Weight:
225.25
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing