Skip to Content
Merck
Dimethyl-1,1,1-d3 sulfide 99 atom % D, 98% (CP)

Dimethyl-1,1,1-d3 sulfide

Dimethyl-1,1,1-d3 sulfide 99 atom % D, 98% (CP)
Synonym(s):
(Methyl sulfide)-1,1,1-d3
Linear Formula:
CH3SCD3
CAS Number:
Molecular Weight:
65.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing