Skip to Content
Merck
DCPIB ≥98% (HPLC)

DCPIB

DCPIB ≥98% (HPLC)
Synonym(s):
4-(2-butyl-6,7-dichloro-2-cyclopentylindan-1-on-5-yl)oxybutyric acid
Empirical Formula (Hill Notation):
C22H28Cl2O4
CAS Number:
Molecular Weight:
427.36

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing