![Daphnetin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/368/666/7a32329e-0fcb-4621-aff5-c540f24d423f/640/7a32329e-0fcb-4621-aff5-c540f24d423f.png)
Daphnetin
![Daphnetin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/368/666/7a32329e-0fcb-4621-aff5-c540f24d423f/640/7a32329e-0fcb-4621-aff5-c540f24d423f.png)
Synonym(s):
7,8-Dihydroxycoumarin, Daphnetin, Daphnetol, Ruixiangsu
Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing