![Cyflufenamid PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/386/753/62c1fe2e-03ab-46db-8d8b-7415df76499f/640/62c1fe2e-03ab-46db-8d8b-7415df76499f.png)
Cyflufenamid
![Cyflufenamid PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/386/753/62c1fe2e-03ab-46db-8d8b-7415df76499f/640/62c1fe2e-03ab-46db-8d8b-7415df76499f.png)
Synonym(s):
(Z)-N-[α-(Cyclopropylmethoxyimino)-2,3-difluoro-6-(trifluoromethyl)benzyl]-2-phenylacetamide
Empirical Formula (Hill Notation):
C20H17F5N2O2
CAS Number:
Molecular Weight:
412.35
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2