
Corning® Axygen® Automation Tips for Zymark®/Caliper

Synonym(s):
Pipette tips for Zymark®/Caliper Automated Liquid Handling System
Products
Product Category
Manufacturer
Color
Package Size
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing