Skip to Content
Merck
cis-3-Hexenyl acetate analytical standard

cis-3-Hexenyl acetate

cis-3-Hexenyl acetate analytical standard
Synonym(s):
(3Z)-3-Hexen-1-ol acetate, (3Z)-C-3-Hexenyl acetate, Leaf acetate
Linear Formula:
CH3CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
142.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3