![Cilostazol Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/333/386/1cdec3e5-ef88-4ea3-a8ec-92fc479968fd/640/1cdec3e5-ef88-4ea3-a8ec-92fc479968fd.png)
Cilostazol Related Compound C
![Cilostazol Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/333/386/1cdec3e5-ef88-4ea3-a8ec-92fc479968fd/640/1cdec3e5-ef88-4ea3-a8ec-92fc479968fd.png)
Synonym(s):
6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butyl]-3,4-dihydro-2(1H)-quinolinone
Empirical Formula (Hill Notation):
C31H45N9O2
CAS Number:
Molecular Weight:
575.75
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2