![Chlortalidone for system suitability, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/223/125/ab8314c8-9f7a-4b1b-81fb-3b1e2ac702dd/640/ab8314c8-9f7a-4b1b-81fb-3b1e2ac702dd.png)
Chlortalidone for system suitability,
![Chlortalidone for system suitability, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/223/125/ab8314c8-9f7a-4b1b-81fb-3b1e2ac702dd/640/ab8314c8-9f7a-4b1b-81fb-3b1e2ac702dd.png)
Synonym(s):
Chlorthalidone
Empirical Formula (Hill Notation):
C14H11ClN2O4S
CAS Number:
Molecular Weight:
338.77
Products
Formula Weight
Gene Alias
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing