Skip to Content
Merck
Chiralyst P461 Umicore

Chiralyst P461

Chiralyst P461 Umicore
Synonym(s):
Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimer, 2,5-Norbornadiene-rhodium(I) chloride dimer, Chloro(2,5-norbornadiene)rhodium(I) dimer, Rhodium(I) chloride 2,5-Norbornadiene complex dimer, [Rh(nbd)Cl]2
Empirical Formula (Hill Notation):
C14H16Cl2Rh2
CAS Number:
Molecular Weight:
460.99
EC Number:

Products

Brand
Formula Weight
Physical Form
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing