Skip to Content
Merck
Carprofen European Pharmacopoeia (EP) Reference Standard

Carprofen

Carprofen European Pharmacopoeia (EP) Reference Standard
Synonym(s):
6-Chloro-α-methyl-9H-carbazole-2-acetic acid, Carprofen
Empirical Formula (Hill Notation):
C15H12ClNO2
CAS Number:
Molecular Weight:
273.71

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique