Skip to Content
Merck
Carmustine United States Pharmacopeia (USP) Reference Standard

Carmustine

Carmustine United States Pharmacopeia (USP) Reference Standard
Synonym(s):
1,3-Bis(2-chloroethyl)-1-nitrosourea, BCNU
Empirical Formula (Hill Notation):
C5H9Cl2N3O2
CAS Number:
Molecular Weight:
214.05

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade