Benztropine mesylate
Synonym(s):
Benzatropine mesylate, Benzotropine mesylate, Benztropine methanesulfonate, Benztropine mesylate
Empirical Formula (Hill Notation):
C21H25NO · CH3SO3H
CAS Number:
Molecular Weight:
403.53
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing