Skip to Content
Merck
Benzaldehyde-DNPH solution certified reference material, 100 μg/mL in acetonitrile (as the aldehyde), ampule of 1 mL

Benzaldehyde-DNPH solution

Benzaldehyde-DNPH solution certified reference material, 100 μg/mL in acetonitrile (as the aldehyde), ampule of 1 mL
Synonym(s):
Benzaldehyde-2,4-dinitrophenylhydrazone solution
CAS Number:

Products

Brand
Product Line
Format
Application
Package Size
Reference Material Type
Special Grade
Technique