Skip to Content
Merck
Bavachin phyproof® Reference Substance

Bavachin

Bavachin phyproof® Reference Substance
Synonym(s):
(2S)-2,3-Dihydro-7-hydroxy-2-(4-hydroxyphenyl)-6-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one, 4′,7-Dihydroxy 6-(3-methyl 2-butenyl)flavanone, Corylifolin
Empirical Formula (Hill Notation):
C20H20O4
CAS Number:
Molecular Weight:
324.37

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing