Aripiprazole
Synonym(s):
7-{4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy}-3,4-dihydro-2(1H)-quinolinone, Aripiprazole
Empirical Formula (Hill Notation):
C23H27Cl2N3O2
CAS Number:
Molecular Weight:
448.39
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4