Skip to Content
Merck
Anti-Thioredoxin–Agarose antibody produced in rabbit IgG fraction of antiserum, PBS suspension

Anti-Thioredoxin–Agarose antibody produced in rabbit

Anti-Thioredoxin–Agarose antibody produced in rabbit IgG fraction of antiserum, PBS suspension
Synonym(s):
Anti-Thioredoxin

Products

Product Category
Brand
Biological Source
Clonality
Physical Form
Analyte Suitability
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1