Anti-γ-Tubulin antibody, Mouse monoclonal
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2