![Angelicin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/157/299/470a3101-5ecf-40f1-8618-020cd7d338ab/640/470a3101-5ecf-40f1-8618-020cd7d338ab.png)
Angelicin
![Angelicin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/157/299/470a3101-5ecf-40f1-8618-020cd7d338ab/640/470a3101-5ecf-40f1-8618-020cd7d338ab.png)
Synonym(s):
2-Oxo-(2H)-furo(2,3-h)-1-benzopyran, 2H-Furo[2,3-h]-1-benzopyran-2-one, Isopsoralen, NSC 404563
Empirical Formula (Hill Notation):
C11H6O3
CAS Number:
Molecular Weight:
186.16
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing