Skip to Content
Merck
Alpha Tocopherol Pharmaceutical Secondary Standard; Certified Reference Material

Alpha Tocopherol

Alpha Tocopherol Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(±)-α-Tocopherol, DL-all-rac-α-Tocopherol, Vitamin E
Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique