![Albuterol certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/416/426/56f9e25b-e57e-4a9a-af86-4ad0fbf9926f/640/56f9e25b-e57e-4a9a-af86-4ad0fbf9926f.png)
Albuterol
![Albuterol certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/416/426/56f9e25b-e57e-4a9a-af86-4ad0fbf9926f/640/56f9e25b-e57e-4a9a-af86-4ad0fbf9926f.png)
Synonym(s):
Salbutamol, α-[(tert-Butylamino)methyl]-4-hydroxy-m-xylene-α,α′-diol, Albuterol
Empirical Formula (Hill Notation):
C13H21NO3
CAS Number:
Molecular Weight:
239.31
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing