Skip to Content
Merck
Adenosine 5′-[β-thio]diphosphate trilithium salt ≥80% (HPLC)

Adenosine 5′-[β-thio]diphosphate trilithium salt

Adenosine 5′-[β-thio]diphosphate trilithium salt ≥80% (HPLC)
Synonym(s):
ADP-β-S, Adenosine-5′-0-(2-thiodiphosphate) trilithium salt, Ado-5′-PP[S]
Empirical Formula (Hill Notation):
C10H12Li3N5O9P2S
CAS Number:
Molecular Weight:
461.07

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing