Skip to Content
Merck
vibrant-m

Acetazolamide Related Compound E

vibrant-m
Synonym(s):
5-Acetamido-1,3,4-thiadiazole-2-sulfonic acid potassium salt, Acetazolamide Related Compound E
Empirical Formula (Hill Notation):
C4H4KN3O4S2
CAS Number:
Molecular Weight:
261.32

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1