Skip to Content
Merck
5-Bromo-2,1,3-benzothiadiazole 95%

5-Bromo-2,1,3-benzothiadiazole

5-Bromo-2,1,3-benzothiadiazole 95%
Synonym(s):
5-Bromobenzo[c][1,2,5]thiadiazole
Empirical Formula (Hill Notation):
C6H3BrN2S
CAS Number:
Molecular Weight:
215.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing