Skip to Content
Merck
4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one analytical standard

4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one

4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one analytical standard
Synonym(s):
Caramel furanone, Sotolon, Sugar lactone
Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing