![2-Fluoro-2-deoxy-D-glucose glycosylation inhibitor, glucose analog](/deepweb/assets/sigmaaldrich/product/structures/265/900/196b7e9b-ba9c-4ddc-a60d-a74b9d7993e1/640/196b7e9b-ba9c-4ddc-a60d-a74b9d7993e1.png)
2-Fluoro-2-deoxy-D-glucose
![2-Fluoro-2-deoxy-D-glucose glycosylation inhibitor, glucose analog](/deepweb/assets/sigmaaldrich/product/structures/265/900/196b7e9b-ba9c-4ddc-a60d-a74b9d7993e1/640/196b7e9b-ba9c-4ddc-a60d-a74b9d7993e1.png)
Synonym(s):
2-Deoxy-2-fluoro-D-glucose, FDG
Empirical Formula (Hill Notation):
C6H11FO5
CAS Number:
Molecular Weight:
182.15
Products
Product Category
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing