![2-(4,6-Diphenyl-1,3,5-triazin-2-yl)-5-[(hexyl)oxy]-phenol analytical standard](/deepweb/assets/sigmaaldrich/product/structures/394/572/1f0f1ab7-38d5-456d-b5cf-409212b6bd6a/640/1f0f1ab7-38d5-456d-b5cf-409212b6bd6a.png)
2-(4,6-Diphenyl-1,3,5-triazin-2-yl)-5-[(hexyl)oxy]-phenol
![2-(4,6-Diphenyl-1,3,5-triazin-2-yl)-5-[(hexyl)oxy]-phenol analytical standard](/deepweb/assets/sigmaaldrich/product/structures/394/572/1f0f1ab7-38d5-456d-b5cf-409212b6bd6a/640/1f0f1ab7-38d5-456d-b5cf-409212b6bd6a.png)
Synonym(s):
2,4-Diphenyl-6-[2-hydroxy-4-(hexyloxy)phenyl]-1,3,5-triazine, 2-(2-Hydroxy-4-hexyloxyphenyl)-4,6-diphenyl-1,3,5-triazine, 2-[4-(Hexyloxy)-2-hydroxyphenyl]-4,6-diphenyl-1,3,5-triazine
Empirical Formula (Hill Notation):
C27H27N3O2
CAS Number:
Molecular Weight:
425.52
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing