Skip to Content
Merck
(−)-2,3-O-Isopropylidene-D-erythronolactone 98%

(−)-2,3-O-Isopropylidene-D-erythronolactone

(−)-2,3-O-Isopropylidene-D-erythronolactone 98%
Empirical Formula (Hill Notation):
C7H10O4
CAS Number:
Molecular Weight:
158.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing