Skip to Content
Merck
2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane 97%

2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane

2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane 97%
Synonym(s):
(2,3,5,6-Tetrafluoro-2,5-cyclohexadiene-1,4-diylidene)dimalononitrile, 7,7,8,8-Tetracyano-2,3,5,6-tetrafluoroquinodimethane, F4TCNQ
Empirical Formula (Hill Notation):
C12F4N4
CAS Number:
Molecular Weight:
276.15

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing