Skip to Content
Merck
2,2′-(Dithiodimethylene)difuran ≥95%, FG

2,2′-(Dithiodimethylene)difuran

2,2′-(Dithiodimethylene)difuran ≥95%, FG
Synonym(s):
Furfuryl disulfide, Difurfuryl disulfide
Empirical Formula (Hill Notation):
C10H10O2S2
CAS Number:
Molecular Weight:
226.32
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing